Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutyltin ≥94%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: TetrabutylstannaneTetra-n-butyltin

93-5049-250G 93-5049-50G
101419-164EA 42.46 USD InStock
101419-164 101419-162
Tetrabutyltin ≥94%
Tetrabutyltin


Formula: C₁₆H₃₆Sn
MW: 347.17 g/mol
Boiling Pt: 145 °C (10 mmHg)
Melting Pt: –97 °C
Density: 1.0572
Flash Pt: 225 °F
Storage Temperature: Ambient
MDL Number: MFCD00009422
CAS Number: 1461-25-2
EINECS: 215-960-8
UN: 1760
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty