Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrapropyltin ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

93-5053-1G 93-5053-5G
100202-182EA 62.28 USD InStock
100202-182 100207-864
Tetrapropyltin ≥97%
Tetrapropyltin


Formula: C₁₂H₂₈Sn
MW: 291.06 g/mol
Boiling Pt: 222 °C
Density: 1.1065 (20°)
MDL Number: MFCD00015196
CAS Number: 2176-98-9
EINECS: 218-536-0
UN: 2788
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty