Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide, anhydrous ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-5616-100G 93-5616-500G
100204-368EA 50.95 USD InStock
100204-368 100197-906
Barium hydroxide, anhydrous ≥95%
Barium hydroxide


Formula: H₂BaO₂
MW: 171.34 g/mol
Melting Pt: 325 °C
Density: 2.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003443
CAS Number: 17194-00-2
EINECS: 241-234-5
UN: 2923
ADR: 8,II
Merck Index: 12,01006
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty