Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum(III) sulfate hydrate ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Dilanthanum trisulfate hydrateDilanthanum trisulphate hydrate

93-5720-50G 93-5720-250G
100204-382EA 329.78 USD InStock
100204-382 100207-878
Lanthanum(III) sulfate hydrate ≥99.9% (La basis)
Lanthanum (III) sulphate hydrate


Formula: La₂O₁₂S₃·H₂O
MW: 584.02 g/mol
Melting Pt: Dec.
Density: 2,821
MDL Number: MFCD00149760
CAS Number: 57804-25-8
EINECS: 233-239-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty