Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tungstic (VI) acid ≥99%

Supplier: STREM CHEMICALS INC MS
Synonyms: Dihydrogen wolframateTungstic acid

93-7410-50G 93-7410-250G
100196-824EA 65.11 USD InStock
100196-824 100210-554
Tungstic (VI) acid ≥99%
Tungstic (VI) acid


Formula: H₂O₄W
MW: 249.85 g/mol
Melting Pt: 100 °C (dec.)
Density: 5,5
MDL Number: MFCD00011336
CAS Number: 7783-03-1
EINECS: 231-975-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty