Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bismuth(III) nitrate pentahydrate ≥98% ACS

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Bismuth trinitrate pentahydrate

93-8304-1000G 93-8304-250G
100210-912EA 163 USD InStock
100210-912 100202-330
Bismuth(III) nitrate pentahydrate ≥98% ACS
Bismuth (III) nitrate pentahydrate


Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 75…80 °C (1013 hPa)
Melting Pt: 30 °C
Density: 2.83
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty