Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony(III) chloride ≥99.999% (Sb basis), PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Antimony trichloride

97-0373-25G
100197-980EA 604.37 USD InStock
100197-980
Antimony(III) chloride ≥99.999% (Sb basis), PURATREM, Electronic Grade
Antimony (III) chloride
Halides for CVD & ALD


Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 283 °C
Melting Pt: 73.4 °C
Density: 3.140
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 13,00713
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty