Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony(III) oxide ≥99.999% (Sb basis), PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: di-Antimony trioxideAntimony trioxideDiantimony trioxide

97-0380-25G 97-0380-100G
100199-676EA 261.85 USD InStock
100199-676 100207-222
Antimony(III) oxide ≥99.999% (Sb basis), PURATREM, Electronic Grade
Antimony (III) oxide


Formula: O₃Sb₂
MW: 291.52 g/mol
Boiling Pt: 1550 °C (subl.)
Melting Pt: 656 °C
Density: 5.67
MDL Number: MFCD00011214
CAS Number: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1,III
Merck Index: 12,00752
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty