Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Paracetamol ≥98%, Powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: APAPParacetamolumAcetaminophenN-(4-Hydroxyphenyl)acetamide4-AcetamidophenolN-Acetyl-4-aminophenol4'-Hydroxyacetanilide

AC100-2.5KG AC100-50KGBL AC100-125GM AC100-500GM
95031-606EA 678 USD InStock
95031-606 95031-608 95031-604 89078-554
Paracetamol ≥98%, Powder USP
Paracetamol
Acetaminophen, Powder, USP, also known as paracetamol, is a widely used antipyretic and mild analgesic that is available over-the-counter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: 168 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5
Merck Index: 13,00048
Order Now
Specification Test Results
Assay 98% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty