Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bismuth chloride oxide ≥79%, powder, purified

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning
Synonyms: Bismuth oxychlorideBismuth(III) chloride oxideBismuth (III) oxychloride

B1118-500GM B1118-25GM B1118-125GM
700001-232EA 163 USD InStock
700001-232 700001-230 700001-234
Bismuth chloride oxide ≥79%, powder, purified
Bismuth chloride oxide
Bismuth Oxychloride, Powder, Purified, also known as bismuthyl chloride, is used as a component in some cosmetics as the primary ingredient included in most skin care powders that are referred to as "mineral makeup. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.


Formula: BiClO
MW: 260.43 g/mol
Density: 7.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010895
CAS Number: 7787-59-9
EINECS: 232-122-7
Order Now
Specification Test Results
Assay 79% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty