Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bronopol ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger
Synonyms: 2-Bromo-2-nitro-1,3-propanediol2-Bromo-2-nitropropane-1,3-diol

B2294-25KGBL
700003-052EA 3522 USD InStock
700003-052
Bronopol ≥98%
Bronopol


Formula: HOCH₂BrC(NO₂)CH₂OH
MW: 199.99 g/mol
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: 130…133 °C
Density: 1.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007390
CAS Number: 52-51-7
EINECS: 200-143-0
UN: 3241
ADR: 4.1,III
Merck Index: 13,01432
Order Now
Specification Test Results
Assay 98% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty