Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y free acid

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Acid Red 872,4,5,7-TetrabromofluoresceinEosin yellowishSolvent red 43BromofluoresceinSpiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-CI 45380Tetrabromofluorescein2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acidBronze Bromo ESBromo acid J. TS, XL, or XX

E1241-25GM
700010-284EA 314.16 USD InStock
700010-284
Eosin Y free acid
Eosin Y
Eosin Y, Free Acid, also known as Solvent Red 43, is used as a personal care colorant.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₂₀H₈Br₄O₅
MW: 647.89 g/mol
Melting Pt: 295 °C
Storage Temperature: Ambient
MDL Number: MFCD00036189
CAS Number: 15086-94-9
EINECS: 239-138-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty