Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oleyl alcohol, Technical Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: cis-Octadec-9-en-1-olcis-9-Octadecen-1-ol

O1021-2.5LT O1021-500ML O1021-200LTB
700002-886EA 290.27 USD InStock
700002-886 700002-884 700002-882
Oleyl alcohol, Technical Grade
Oleyl alcohol
Oleyl Alcohol, Technical, also known as Octadecenol, is a fatty alcohol from beef fat that is used as a nonionic surfactant, emollient and emulsifier.

Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.


Formula: C₁₈H₃₆O
MW: 268.48 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 13…19 °C
Density: 0.849
Storage Temperature: Ambient
MDL Number: MFCD00002993
CAS Number: 143-28-2
EINECS: 205-597-3
Merck Index: 12,06968
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty