Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium decyl sulfate ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Decyl sodium sulphateDecyl sulfuric acid saltDecyl sodium sulfateDecyl sulphuric acid salt

S1259-25GM
700006-714EA 706 USD InStock
700006-714
Sodium decyl sulfate ≥95%
Sodium decyl sulphate
Sodium Decyl Sulfate is an anionic surfactant with medium foaming, low viscosity and low cloud point profile and is a superior dispersing, wetting and emulsifying agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₁₀H₂₁NaO₄S
MW: 260.33 g/mol
Melting Pt: 191.85 °C
Density: 1.207 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041881
CAS Number: 142-87-0
EINECS: 205-568-5
Order Now
Specification Test Results
Assay 95% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty