Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(IV) oxide ≥99.9%, powder, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Tin dioxideStannic OxideTin(iv)oxide

S1530-25GM S1530-2.5KG S1530-125GM S1530-45KGBL S1530-12KG S1530-500GM
700000-940EA 5817 USD InStock
700000-940 700000-942 700000-932 700000-934 700000-936 700000-938
Tin(IV) oxide ≥99.9%, powder, Reagent Grade
Tin (IV) oxide
Stannic Oxide, Powder, Reagent is also known as Tin(IV) oxide and is an inorganic compound and along with vanadium oxide, is used in the oxidation of aromatic compounds in the synthesis of acid anhydrides and carboxylic acids. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.


Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: ∼2500 °C (1013 hPa)
Melting Pt: 1624.85 °C
Density: 6.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,08778
Order Now
Specification Test Results
Assay 99.9% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty