Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene-4-sulfonamide ≥97.5%

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 4-Methylbenzenesulphonamide4-Methylbenzenesulfonamidep-Toluenesulfonamidep-Toluenesulphonamide

T1310-1KG T1310-250GM
700009-298EA 139 USD InStock
700009-298 700009-300
Toluene-4-sulfonamide ≥97.5%
Toluene-4-sulphonamide
p-Toluenesulfonamide is used as synthetic intermediate and as a film forming agent in cosmetics.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00011692
CAS Number: 70-55-3
EINECS: 200-741-1
Order Now
Specification Test Results
Assay 97.5% Min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty