Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Serine

Supplier: R&D Systems
Synonyms: (R)-(-)-SerineH-D-Ser-OH

0226/1G
100505-382EA 183.66 USD InStock
100505-382
D-(-)-Serine
D(-)-Serine


Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 235 °C
MDL Number: MFCD00004269
CAS Number: 312-84-5
EINECS: 206-229-4
Merck Index: 12,08604
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty