Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminobutyric acid

Supplier: R&D Systems
Synonyms: γ-Aminobutyric acidGABA

0344/1G
100505-100EA 120.03 USD InStock
100505-100
4-Aminobutyric acid
4-Aminobutyric acid


Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 202 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty