Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-2,4,6-triiodobenzoic acid ≥98%

Supplier: USP
Warning

1026605
101096-782EA 729.23 USD InStock
101096-782
3-Amino-2,4,6-triiodobenzoic acid ≥98%
3-Amino-2,4,6-triiodobenzoic acid


Formula: C₇H₄I₃NO₂
MW: 514.83 g/mol
Boiling Pt: 459 °C (1013 hPa)
Melting Pt: 196 °C
MDL Number: MFCD00007681
CAS Number: 3119-15-1
EINECS: 221-493-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty