Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Acetylsalicylic acid

Supplier: USP
Warning
Synonyms: AspirinAspirin®2-Acetoxybenzoic acidAcetylsalicylic acid

1044006
101094-328EA 420 USD InStock
101094-328
O-Acetylsalicylic acid
o-Acetylsalicylic acid


Formula: 2-(CH₃CO₂)C₆H₄CO₂H
MW: 180.16 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 138…140 °C
Density: 1.39 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00002430
CAS Number: 50-78-2
EINECS: 200-064-1
Merck Index: 13,00856
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/25/2296925.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty