Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Biotin

Supplier: USP
Synonyms: Vitamin HVitamin B85-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acidD-BiotinVitamin B7

1071508
101094-408EA 429.23 USD InStock
101094-408
D-(+)-Biotin
D(+)-Biotin


Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Melting Pt: 232…233 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty