Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Cetylpyridinium chloride monohydrate

Supplier: USP
Danger
Synonyms: Hexadecylpyridinium chloride monohydrate1-Hexadecylpyridinium chloride monohydrateCetylpyridinium Chloride, Monohydrate

1104006
101094-574EA 407.69 USD InStock
101094-574
1-Cetylpyridinium chloride monohydrate
1-Cetylpyridinium chloride monohydrate


Formula: C₂₁H₃₈ClN·1H₂O
MW: 358.01 g/mol
Melting Pt: 77…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149977
CAS Number: 6004-24-6
EINECS: 204-593-9
UN: 2811
ADR: 6.1,I
Merck Index: 13,02041
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty