Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cholecalciferol (Vitamin D3)

Supplier: USP
Synonyms: Vitamin D3Colecalciferol

1131009
101096-454EA 436.92 USD InStock
101096-454
Cholecalciferol (Vitamin D3)
Cholecalciferol (Vitamin D3)


Formula: C₂₇H₄₄O
MW: 384.65 g/mol
MDL Number: MFCD00078131
CAS Number: 67-97-0
EINECS: 200-673-2
UN: 2811
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty