Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clioquinol

Supplier: USP
Danger
Synonyms: 5-Chloro-8-hydroxy-7-iodoquinolineClioquinol5-Chloro-7-iodo-8-quinolinol

1138201
101094-648EA 427.69 USD InStock
101094-648
Clioquinol
Clioquinol


Formula: C₉H₅ClINO
MW: 305.5 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 183…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006787
CAS Number: 130-26-7
EINECS: 204-984-4
UN: 2811
ADR: 6.1,III
Merck Index: 12,05052
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty