Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dexamethasone-21-acetate

Supplier: USP
Synonyms: 11β,17α-Dihydroxy-21-(ethanoyloxy)-9-fluor-16α-methyl-1,4-pregnadien-3,20-dionDexamethasone acetate

1176506
101094-754EA 420 USD InStock
101094-754
Dexamethasone-21-acetate
Dexamethasone 21-acetate


Formula: C₂₄H₃₁FO₆
MW: 434.5 g/mol
MDL Number: MFCD00027407
CAS Number: 1177-87-3
EINECS: 214-646-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty