Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Diethyl-m-toluamide

Supplier: USP
Warning
Synonyms: DiethyltoluamideDeetDiethyl-m-toluamideN,N-Diethyl-3-methylbenzamide

1197007
101094-098EA 420 USD InStock
101094-098
N,N-Diethyl-m-toluamide
N,N-Diethyl-m-toluamide


Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0,996 g/cm³ (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00009046
CAS Number: 134-62-3
EINECS: 205-149-7
Merck Index: 13,02876
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty