Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluorouracil

Supplier: USP
Danger
Synonyms: 5-Fluoropyrimidine-2,4(1H,3H)-dioneFluorouracil5-FU

1279000
101095-036EA 420 USD InStock
101095-036
5-Fluorouracil
5-Fluorouracil


Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 282…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 12,04219
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty