Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hexylresorcinol

Supplier: USP
Warning
Synonyms: 4-Hexylbenzene-1,3-diol

1308307
101095-130EA 401.54 USD InStock
101095-130
4-Hexylresorcinol
4-Hexylresorcinol


Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 64…67 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002284
CAS Number: 136-77-6
EINECS: 205-257-4
Merck Index: 12,04750
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty