Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminosalicylic acid

Supplier: USP
Synonyms: 5-Amino-2-hydroxybenzoic acidMesalamineMesalazinem-Aminosalicylic acid

1392705
101095-436EA 420 USD InStock
101095-436
5-Aminosalicylic acid
5-Aminosalicylic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 280 °C
Density: 1.568 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007877
CAS Number: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty