Would you like to log in again?
Reference Electrodes, Hach
Supplier: Hach
B20C820 B20B300 LZW5044970002 E21M003
89174-316EA 2311.22 USD InStock
89174-316 89174-228 89174-312 89174-314
Reference Electrodes, Hach
Electrodes pH Electrodes
A reference electrode provides a stable potential whatever the measurement conditions.
These reference electrodes have a glass body and are available with a variety of lengths, diameters and reference systems to meet your analysis needs. The distinguishing characteristics between reference electrodes are the type of reference system and the liquid junction.
Certifications: CE.WEEE.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty