Would you like to log in again?
PadMop™ Covered Sponge Mops, Micronova
Supplier: Micronova
PadMop™
SLP6-18 SLP6-14
68400-702CS 447.74 USD InStock
68400-702 68400-700
PadMop™ Covered Sponge Mops, Micronova
Mops
This fully padded, brick-style flat mop is designed to apply generous amounts of disinfectant to cope with railings, shelving, door jambs, corners, and large surface areas in pharmaceutical production areas.
  • Dual padded cleaning surfaces for railings, door jambs, and corners
  • 0.75" (19.05 mm) urethane foam interiors
  • Holds up to common disinfectants and cleaning agents
  • Launderable and reusable
  • Autoclavable
Fully encased in a choice of low-particle fabric:
NovaPoly - 100% smooth knit polyester provides excellent disinfectant dispersal.
PolySorb - 100% textured polyester for removing residues and dispersing disinfectant.
The Micronova PadMop™ Covered Sponge Mop features dual cleaning surfaces for generous dispersement of solution on primary surfaces as well as using the top of the pad for cleaning door jams, railings, piping, and more. The PadMop™ fits onto Micronova’s 360° swivel SlimLine™ adapter, or the LyoTool™ for cleaning lyophilizer shelving.
Ordering information: All components must be ordered separately. For additional handle lengths, see Universal Mop Handles or Plastic Mop Handles from Micronova.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty