Would you like to log in again?
InGel™ Blue for Protein Digestion, G-Biosciences
Supplier: G-Biosciences
786-681 786-681
89167-892EA 330.03 USD InStock
CA89167-892 89167-892
InGel™ Blue for Protein Digestion, G-Biosciences
Protein Extraction Protein Extraction Kits
G-Biosciences' InGel™ Blue kit provides a complete set of reagents for the in gel tryptic digestion and extraction of peptides for mass spectrometry (MALDI and LC MS/MS)
The protein spots are first excised from the Coomassie or fluorescent stained gel. The stained gel pieces are washed with G-Biosciences' BlueOUT™ to remove inhibitory stains. The protein is then alkylated and reduced within the gel piece using the supplied aliquots of OneQuant™ DTT and OneQuant™ Iodoacetamide.
The proteins are digested within the gel using our Mass Spectrometry Grade Trypsin and proprietary Digestion Buffer. The digested peptides are extracted with Pep-Extract™, a high diffusion peptide extraction buffer. The extracted peptides are suitable for mass spectrometry analysis without any subsequent treatments or cleaning procedures. The InGel™ Blue kit is suitable for about 100 protein spots or bands.
G-Biosciences' InGel™ Blue kit is supplied with BlueOUT™ for the removal of Coomassie or fluroescent stains, OneQuant™ DTT used as a reducing agent and supplied in single use aliquots to prevent contamination, OneQuant™ Iodoacetamide used as an alkylating agent, again in single use aliquots to prevent contamination, Mass Spectrometry Grade Trypsin, Trypsin Digestion Buffer, and Pep-Extract™ for high level peptide extraction.
The kit is specifically designed for use with Coomassie or fluorescent stained protein spots/bands.
Ordering information: Accessories included.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty