Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Advanced TC™ Treated Cell Culture Dishes, Vented, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
664960 639960
89131-700CS 865.08 USD InStock
89131-700 89501-636
Advanced TC™ Treated Cell Culture Dishes, Vented, Sterile, Greiner Bio-One
Culture Dishes Cell Culture Dishes
Advanced Tissue Culture (TC) treated Cell Culture Dishes, made from virgin polystyrene, allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
  • Virgin polystyrene construction
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Sterile
Advanced TC™ surface treatments provide the conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield.
Advanced TC™ Cell Culture Dishes can be used with serum-reduced or serum-free media, transfection, transduction and high througput analysis. Dishes are vented for optimal gas exchange.
Sterile, DNase- Free, RNase- Free, human DNA - Free, nonpyrogenic and noncytotoxic. Store at room temperature. Two year shelf life.
Ordering information: Sterile and user friendly packaging. Available with clear lid.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty