Would you like to log in again?
Concept Motorized Modular Workstations
Supplier: Treston
10335132 10335133 10335131
89202-098EA 4003.12 USD InStock
89202-098 89202-100 89202-102
Concept Motorized Modular Workstations
Furniture Tables and Workbenches
Concept lab benches are particularly suitable for the needs of laboratory industries.
  • Desk switch can be installed on the front, left or right side of the frame
  • The Concept lab bench adapts to the work process and the needs of the user
Concept motor frames can support up to 1100 lbs. static, or 660 lbs. dynamic. The speed of the frame is 0.7"/sec without load, or 0.59"/sec with load.
The desk switch has a multi position memory system to be able to set the frame to three customer based heights and up-down buttons for random settings.
There are many possible adjustments, all components fit together seamlessly, and each detail is practical in use. All movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. Concept lab benches are also available in handcrank and friction versions. Upright frames can be added to allow for use of accessories. Height adjustable from 54.8 to 48.3".
Ordering information: Work surfaces sold separately. Corner, extension, and double-sided table add on options available. Installation is required. Assembly videos available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty