Would you like to log in again?
FASTsilver™, Rapid Silver Stain for Protein Gels and Nucleic Acids, G-Biosciences
Supplier: G-Biosciences
FASTsilver™
786-30 786-30
82021-392EA 293.2 USD InStock
82021-392 CA82021-392
FASTsilver™, Rapid Silver Stain for Protein Gels and Nucleic Acids, G-Biosciences
Stains Electrophoresis Stains
G-Biosciences' FASTsilver™ is a rapid silver stain for nucleic acids and proteins in electrophoresis gels
  • Nanogram-sensitive
  • Stains both nucleic acids & proteins
  • Maximum visibility: produces crystal clear backgrounds & sharp protein bands
  • Lower Detection Limit: 1ng/band
FASTsilver™ does not use the protein modifier glutaraldehyde, and therefore, it allows for complete recovery of proteins and trypsin digested peptides.
G-Biosciences' FASTsilver™ detects as little as 0.3 ng nucleic acid and 1 ng BSA protein. The kit contains ready-to-use reagents for 25 mini-gels and comes with a simple-to-follow 60 to 90 minute protocol.
The FASTsilver™ kit is a nanogram-sensitive silver staining kit that produces crystal clear background and maximal sensitivity needed for critical analysis. This unique formulation of reagents leaves the background clear and produces sharp images of nucleic acid bands.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty