Would you like to log in again?
FOCUS™ SubCell, G-Biosciences
Supplier: G-Biosciences
786-260
82023-268EA 642.42 USD InStock
CA82023-268 82023-268
FOCUS™ SubCell, G-Biosciences
Protein Extraction Protein Extraction Kits
G-Biosciences' FOCUS™ SubCell is for the total subcellular fractionation of cells and mammalian tissue into enriched fractions of nuclear, mitochondrial, cytosolic and membrane proteins.
The resulting mitochondrial fraction can be subsequently separated into heavy and light fractions by gradient centrifugation and the resulting mitochondria are >90% active and have intact inner and outer membranes.
The kit includes reagents for optional steps that minimize contaminations of the nuclear fraction by cytoplasmic elements.
Suitable for cultured animal cells and adaptable for animal tissues. Designed for 50 preps, where one prep is equivalent to 2x10^7 mammalian cells or 100 mg mammalian tissue.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty