Would you like to log in again?
Fraction-FOCUS™, G-Biosciences
Supplier: G-Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
786-168
82022-578PK 804.33 USD InStock
CA82022-578 82022-578
Fraction-FOCUS™, G-Biosciences
Protein Extraction Protein Extraction Kits
2D electrophoresis and mass spectrometry is routinely used for identification of novel proteins, however the greatest challenge in protein identification is achieving suitable resolution of proteins
Fractionation simplifies the protein composition and allows for improved resolution and simplified 2D maps, which in turn allows for improved analysis and interpretation and greatly increases the chances of identifying novel and less abundant proteins.
G-Biosciences' Fraction-FOCUS™ utilizes proven technology to fractionate and concentrate all proteomes into multiple fractions; simplifying 2D maps and enhancing detection of low abundant proteins. Fraction-FOCUS™ is fully compatible with all downstream protein identification techniques.
There is no detectable loss of total protein during the Fraction-FOCUS™ procedure. At the end of the fractionation, cellular proteins are in one of many fractions.
The high dynamic range of a species’ proteome means that the more abundant proteins mask the less abundant and often more interesting proteins.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty