Would you like to log in again?
Nunc® CryoRacks for CryoTubes, Thermo Scientific
Supplier: Thermo Scientific
367000 367000
73521-346CS 583.05 USD InStock
73521-346 CA73521-346
Nunc® CryoRacks for CryoTubes, Thermo Scientific
Racks Cryogenic Box Racks
Nunc® CryoRacks for CryoTubes™ fit most chest freezers, have adjustable shelves to fit many box sizes, have alignment rods to secure boxes in position and are easy-to-clean.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty