Would you like to log in again?
Eppendorf® Eporator® Electroporation System
4309000027 4309000027
71002-390EA 3740 USD InStock
CA11007-356 71002-390
Eppendorf® Eporator® Electroporation System
Electroporation Systems Electroporators
The Eporator® is a compact instrument designed for fast and controlled electroporation of bacteria, yeast, and other microorganisms
The unit ensures faster sample handling due to simple, intuitive operation. Two program buttons allow storage and recall of most commonly used parameters. Safe electronics and an integrated electroporation chamber eliminate voltage leaks and misuse. The compact, space-saving design allows for easy transport and storage. GLP-compliant documentation is quckly and easily made available via USB port.
The instrument is unique due to its intuitive operation and user-friendly programming of standard methods, as well as its USB port where experimental data may be easily exported and documented.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty