Would you like to log in again?
Eppendorf® twin.tec real-time PCR Plates
Supplier: Eppendorf
951022043 951022015 0030132718 951022015 30132742 951022003 0030132718 951022043 0030132700 951022055 951022003 951022055 0030132700
89136-716PK 161.53 USD InStock
89136-716 CA89136-714 89087-442 89176-456 89136-714 CA89087-444 CA89136-716 CA89087-442 89087-444 89087-438 CA89087-436 89087-436 CA89087-438
Eppendorf® twin.tec real-time PCR Plates
Microplates PCR Plates
Eppendorf® twin.tec real-time PCR Plates offer all of the advantages of regular twin.tec plates with the advantage of white wells for real-time PCR applications
  • Plates are available in skirted, semi-skirted, or unskirted frames
  • 96- or 384-well formats available
  • Optimal heat transfer due to reduced wall thickness
  • Plates offer high mechanical stability
  • Raised well rims ensure effective sealing
  • Autoclavable at 121°C for 20 minutes
  • Available with barcode (upon request)
White wells significantly reduce interfering background fluorescence, and lead to increased homogeneity of replicates and reproducible results. These plates combine extremely thin walled polypropylene wells for optimal heat transfer to the sample and extremely robust polycarbonate frames for ultimate rigidity and torque-resistance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty