Would you like to log in again?
Heat Sealing PCR Film and Foil, Eppendorf®
Supplier: Eppendorf
0030127838 0030127854 0030127838
75875-058CS 293.39 USD InStock
75875-058 CA89167-286 CA89167-284
Heat Sealing PCR Film and Foil, Eppendorf®
Seals Microplate Seals
Eppendorf® heat sealing film and foil are designed for hermetic sealing of multiwell plates, particularly in low volume reactions.
Both film and foil effectively prevent evaporation loss during PCR reactions. The optically clear polyester/polypropylene laminate film is an extremely stable sealing option that resists removal or piercing. The laminated aluminum foil is easily pierceable and removeable. Both film and foil are certified free of human DNA, DNase, RNase and PCR inhibitors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty