Would you like to log in again?
Tantalum Boat, Thermo Scientific
Supplier: Thermo Scientific
041217-KU
AAA41217-KUEA 123.99 USD InStock
AAA41217-KU
Tantalum Boat, Thermo Scientific
Boats Combustion Boats
Tantalum Boat is designed for disc aperture cleaning and vacuum evaporation applications.
Store at ambient temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty