Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride for spectroscopy

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: HaliteSea saltSaline

43631.19 43631-19
AAA43631-19EA 158.99 USD InStock
CAAAA43631-19 AAA43631-19
Sodium chloride for spectroscopy
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C
Melting Pt: 801 °C
Density: 2.165
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty