Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(IV) oxide 15% in water, colloidal dispersion

Supplier: Thermo Scientific Chemicals
Synonyms: Tin dioxideStannic OxideTin(iv)oxide

44592-A3 44592.36 044592.A3 44592-36
AAA44592-36EA 93 USD InStock
AAA44592-36 AAA44592-A3 CAAAA44592-36 CAAAA44592-A3
Tin(IV) oxide 15% in water, colloidal dispersion
Tin (IV) oxide


Formula: O₂Sn
MW: 150.71 g/mol
Density: 1.153
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty