Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bismuth(III) oxide ≥99.5%, NanoArc®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dibismuth trioxideBismuth Trioxide

45582-36 45582-14 45582-22
AA45582-22EA 128.99 USD InStock
AA45582-22 AA45582-36 AA45582-14
Bismuth(III) oxide ≥99.5%, NanoArc®
Bismuth (III) oxide
X-ray opaque particle. NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties.


Formula: Bi₂O₃
MW: 465.96 g/mol
Boiling Pt: ∼1890 °C
Melting Pt: 817 °C
Density: 8,9
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty