Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A11925-0B A11925-30 A11925-30 A11925-18 A11925-18 A11925-0B
AAAA11925-18EA 77.79 USD InStock
AAAA11925-18 AAAA11925-30 CAAAAA11925-30 AAAA11925-0B CAAAAA11925-0B CAAAAA11925-18
Iodic acid 99%
Iodic acid


Formula: HIO₃
MW: 175.91 g/mol
Melting Pt: 110 °C (dec.)
Density: 4.629
Storage Temperature: Ambient
MDL Number: MFCD00011348
CAS Number: 7782-68-5
EINECS: 231-962-1
UN: 3085
ADR: "5.1",III
Merck Index: 13,05034
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty