Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A16029-06 A16029-14 A16029-03 A16029-06 A16029-03 A16029-14
AAAA16029-06EA 80.34 USD InStock
AAAA16029-06 AAA16029-03 CAAAA16029-03 CAAAAA16029-14 AAAA16029-14 CAAAAA16029-06
4-Chloro-3-fluorophenol 98%
4-Chloro-3-fluorophenol


Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 84 °C/44 mm
Melting Pt: 50…54 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042583
CAS Number: 348-60-7
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty