Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H26092-MD H26092-03
AAH26092-MDEA 58.41 USD InStock
AAH26092-MD AAH26092-03
4-Amino-3-fluorobenzoic acid 98%
4-Amino-3-fluorobenzoic acid


Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD01660374
CAS Number: 455-87-8
EINECS: 207-250-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty