Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biuret 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Allophanic acid amideCarbamylurea

L00812-36 L00812-22
AAAL00812-22EA 72.75 USD InStock
AAAL00812-22 AAAL00812-36
Biuret 97%
Biuret
Biuret is used in protein assay. It is also used as Catalytic agent, Petrochemical additive. It is used in organic synthesis. Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.


Formula: C₂H₅N₃O₂
MW: 103.08 g/mol
Melting Pt: ca 189 °C (dec.)
Density: 1.46
Storage Temperature: Ambient
MDL Number: MFCD00007946
CAS Number: 108-19-0
EINECS: 203-559-0
Merck Index: 13,01305
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/26/2353326.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty