Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentadecanol 99%

Supplier: Thermo Scientific Chemicals

L03673-06 L03673-06
AAAL03673-06EA 85.2 USD InStock
AAAL03673-06 CAAAAL03673-06
1-Pentadecanol 99%
1-Pentadecanol


Formula: H₃C(CH₂)₁₃CH₂OH
MW: 228.42 g/mol
Boiling Pt: 269…271 °C
Melting Pt: 45…46 °C
Density: 0.893 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004759
CAS Number: 629-76-5
EINECS: 211-107-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty